EMAIL THIS PAGE TO A FRIEND
Meditsinskaia parazitologiia i parazitarnye bolezni
[The results of a trial of the insecticidal activity of Oradelt and Omait against rodent fleas].
PMID 7770015
Abstract
The paper provides the results of examining the insecticidal activity of the commercial agents oradelt and omait against rodent fleas. The two agents have been ascertained to have a high toxicity for fleas of various species and genera. The field trials have shown that oradelt has a dusticidal effect in the habitats of mountain suslik. The oradelt dust containing 0.05% of the active ingredient should be recommended for controlling fleas in the foci of the suslik-type plague.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32051
Propargite, PESTANAL®, analytical standard, technical mixture
C19H26O4S