EMAIL THIS PAGE TO A FRIEND
Nature genetics
Mutation of the MXI1 gene in prostate cancer.
PMID 7773287
Abstract
The Mxi1 protein negatively regulates Myc oncoprotein activity and thus potentially serves a tumour suppressor function. MXI1 maps to chromosome 10q24-q25, a region that is deleted in some cases of prostate cancer. We have detected mutations in the retained MXI1 alleles in four primary prostate tumours with 10q24-q25 deletions. Two tumours contained inactivating mutations, whereas two others contained the identical missense mutation. Fluorescence in situ hybridization also demonstrated loss of one MXI1 allele in an additional tumour lacking chromosome 10 abnormalities. MXI1 thus displays allelic loss and mutation in some cases of prostate cancer that may contribute to the pathogenesis or neoplastic evolution of this common malignancy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV31403
Anti-MXI1 antibody produced in rabbit, affinity isolated antibody