EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Allergic contact dermatitis from tolnaftate.
PMID 7774192
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T6638
Tolnaftate
C19H17NOS