EMAIL THIS PAGE TO A FRIEND
The Journal of cell biology
The interaction of the retina cell surface N-acetylgalactosaminylphosphotransferase with an endogenous proteoglycan ligand results in inhibition of cadherin-mediated adhesion.
PMID 7775582