EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Enzymatic synthesis of guanine nucleotides labeled with 15N at the 2-amino group of the purine ring.
PMID 7778777
Abstract
GMP and dGMP labeled with 15N at the 2-amino group of the purine ring was obtained enzymatically from NH4Cl (> 99 at.% 15N) and from IMP or dIMP, respectively, by several reactions involving IMP-dehydrogenase, GMP-synthetase, adenylate kinase, and creatine kinase. The first three enzymes were obtained by overexpression in Escherichia coli of the corresponding genes. The isotope content of the primary amino group of guanine determined by mass spectrometry after acid hydrolysis of nucleotides was found higher than 98 at.% 15N. The proton NMR spectrum of [15N]GMP in solution in the absence of nitrogen decoupling showed a doublet with a coupling constant of 92 Hz. When nitrogen decoupling was used during the acquisition time, the doublet was replaced by a single peak at 6.47 ppm, indicating that the corresponding proton is bound to 15N.