EMAIL THIS PAGE TO A FRIEND
Journal of applied toxicology : JAT
Acute renal and hepatic effects induced by 3-haloanilines in the Fischer 344 rat.
PMID 7782560
Abstract
Haloanilines are commonly used as chemical intermediates in the manufacture of a wide range of products. The purpose of this study was to examine the in vivo nephrotoxic and hepatotoxic potentials of the 3-haloanilines. The in vitro effects of the 3-haloanilines on renal function were also examined. In the in vivo experiments, male Fischer 344 rats (four rats/group) were administered a single intraperitoneal (i.p.) injection of an aniline hydrochloride (1.0 or 1.25 mmol kg-1) or vehicle. Renal and hepatic function were monitored at 24 and/or 48 h post-treatment. None of the 3-haloanilines were potent nephrotoxicants at either dose level. The greatest effects on renal function were observed following administration of 3-chloroaniline at a dose of 1.25 mmol kg-1 (oliguria, glucosuria, hematuria, decreased p-aminohippurate accumulation by renal cortical slices and increased blood urea nitrogen concentration). 3-Chloroaniline also was the only aniline compound to increase plasma ALT/GPT activity at 48 h. In the in vitro experiments, the ability of an aniline (10(-5) - 10(-3) M) to decrease organic ion accumulation in renal cortical slices from untreated rats was examined. The decreasing order of in vitro nephrotoxic potential was 3-iodoaniline > 3-bromoaniline > 3-chloroaniline > aniline > 3-fluoroaniline. These results indicate that the 3-haloanilines are not potent nephrotoxicants or hepatotoxicants at sublethal doses. In addition, the reasons why the 3-haloanilines have different orders of nephrotoxic potential in vivo and in vitro are not clear at this time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
180025
3-Bromoaniline, 98%
C6H6BrN