EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Molecular cloning of a cDNA for proctase B from Aspergillus niger var. macrosporus and sequence comparison with other aspergillopepsins I.
PMID 7787314
Abstract
A cDNA for proctase B from Aspergillus niger var. macrosporus was isolated and sequenced. The deduced amino acid sequence (394 residues) of the preproform of the enzyme was highly homologous (98% identify) with those of aspergillopepsins I from A. awamori and A. saitoi, and moderately homologous (68% identity) with that of A. oryzae. Most of the sequence differences were found in the carboxyl-terminal domain.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P2143 Protease from Aspergillus saitoi, Type XIII, ≥0.6 unit/mg solid