EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Binding and modification of proteins by methylglyoxal under physiological conditions. A kinetic and mechanistic study with N alpha-acetylarginine, N alpha-acetylcysteine, and N alpha-acetyllysine, and bovine serum albumin.
PMID 7798230
Abstract
The physiological alpha-oxoaldehyde methylglyoxal binds and modifies arginine, lysine, and cysteine residues in proteins. The kinetics and mechanism of these reactions were investigated with N alpha-acetylamino acids and bovine serum albumin at pH 7.4 and 37 degrees C. The reaction of methylglyoxal with N alpha-acetylarginine involved the initial reversible formation of glycosylamine and 4,5-dihydroxy-5-methylimidazolidine derivatives, with further slow irreversible conversion to an imidazolone, N alpha-acetyl-N delta- (5-methyl-4-imidazolon-2-yl)ornithine. The imidazolone was fluorescent with an excitation lambda max value of 320 nm and an emission lambda max value of 398 nm. Methylglyoxal reacted reversibly with N alpha-acetyllysine to form glycosylamine and bisglycosylamine derivatives. Further reaction of these glycosylamines occurred to form brown, fluorescent oligomers that were not characterized. Methylglyoxal reacted rapidly and reversibly with N alpha-acetylcysteine to form the hemithioacetal adduct. The reaction of methylglyoxal with bovine serum albumin (BSA) at pH 7.4 and 37 degrees C involved the reversible and irreversible formation of methylglyoxal-BSA adducts. Irreversible modification of BSA occurred mainly on arginine residues to form imidazolone. The formation of methylglyoxal-modified proteins involves glycoxidation leading to advanced glycation end product-like fluorescence. It is expected to be increased in diabetes mellitus and may be linked to the development of diabetic complications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3133
Nα-Acetyl-L-arginine
C8H16N4O3