EMAIL THIS PAGE TO A FRIEND
The Journal of pharmacy and pharmacology
The inhibitory effects of cephalosporin and dipeptide on ceftibuten uptake by human and rat intestinal brush-border membrane vesicles.
PMID 7815284
Abstract
The types of inhibitory effects caused by compound V (an analogue of ceftibuten) and alanylproline (dipeptide) on the uptake of ceftibuten by brush-border membrane vesicles (BBMV) prepared from human and rat small intestine were analysed. In the presence of an inward H(+)-gradient, the initial uptake rate of ceftibuten by both human and rat intestinal BBMV was concentration-dependent with apparent Km and Vmax values of 0.35 mM and 2.052 nmol (mg protein)-1 min-1 for human BBMV, and 0.50 mM and 3.056 nmol (mg protein)-1 min-1 for rat BBMV, respectively. For both human and rat BBMV, kinetic analysis by Dixon and Lineweaver-Burk plots demonstrated that the uptake of ceftibuten was competitively inhibited by compound V, whereas inhibition by alanylproline was noncompetitive or partially competitive. These results suggest that there is a stereospecific transport system which is common to ceftibuten and compound V, and that this system is not identical to the carrier system for the dipeptide, alanylproline.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3253
Ala-Pro hydrate
C8H14N2O3 · xH2O