EMAIL THIS PAGE TO A FRIEND
Minerva stomatologica
[An in-vitro study of the antimicrobial activity of copper and zinc salts on pure and mixed microbial cultures].
PMID 7816012
Abstract
The authors evaluated the in vitro antimicrobial activity of some copper and zinc salts. The results showed high antimicrobial activity of copper salts versus S. mutans and versus the bacterial association tested in this study.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
205567
Zinc fluoride, powder, 99%
F2Zn