EMAIL THIS PAGE TO A FRIEND
Nucleic Acids Research
Polymerase chain reaction products containing 5-methyldeoxycytidine: a microplate immunoquantitation method.
PMID 7816646
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MABE146
Anti-5-methylcytosine Antibody, clone 33D3, clone 33D3, from mouse