EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Axial skeleton homeosis and forelimb malformations in Hoxd-11 mutant mice.
PMID 7816839
Abstract
The Hoxd-11 gene was disrupted by homologous recombination in embryonic stem cells. We found that Hoxd-11-/- mutant mice are viable and display homeotic transformations of their sacral vertebrae, while their forelimbs present abnormalities of some metacarpals and of the first row of carpal bones. These results are discussed in the light of current models of tetrapod axial skeleton and limb patterning.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV31966
Anti-HOXD11 antibody produced in rabbit, affinity isolated antibody