EMAIL THIS PAGE TO A FRIEND
Australian veterinary journal
The efficacy of a combination anthelmintic against oxibendazole resistant small strongyles, large strongyles and ascarids in horses.
PMID 7818443