EMAIL THIS PAGE TO A FRIEND
Journal de gynecologie, obstetrique et biologie de la reproduction
[Non-steroidal anti-inflammatory agents and pregnancy. A study of renal and digestive toxicity of niflumic acid in the perinatal period].
PMID 7822716
Abstract
We report a case of intrauterine exposure to niflumic acid in a preterm neonate (35 weeks of gestation). The mother received niflumic acid (750 mg daily) during the last four days of gestation. Severe oligohydramnios was present. A transient neonatal renal failure, but also abdominal complications were observed for several days. Maternal and neonate niflumic acid levels were studied and showed an important placental transfer. Niflumic acid is a prostaglandin synthetase inhibitor. It is important that the nephrotoxicity of this vasoactive drug be known. Maternal administration of niflumic acid during the last days of gestation can induce fetal and neonatal adverse effects, especially renal failure. Prescription of all nonsteroidal anti-inflammatory drugs must be very cautious during pregnancy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N0630
Niflumic acid
C13H9F3N2O2
Y0000824
Niflumic acid, European Pharmacopoeia (EP) Reference Standard
C13H9F3N2O2