EMAIL THIS PAGE TO A FRIEND
Current opinion in oncology
Systemic administration of new therapeutic radioisotopes, including phosphorus, strontium, samarium, and rhenium.
PMID 7827173
Abstract
New advances in systemic radionuclide therapy have increased the number of treatment options available for patients with painful osseous metastases. This form of therapy has three major appeals: 1) it addresses all sites of involvement; 2) selective absorption into bone limits irradiation of normal tissues; and 3) as a result, toxicity may be reduced and the therapeutic ratio improved. The clinical experience with radioactive phosphorus, strontium, samarium, and rhenium are reviewed. To date, the best studied and the only Food and Drug Administration approved agent is strontium-89. About 60% to 90% of patients treated with strontium-89 respond with complete or partial relief of pain for a median duration of 6 months. Large, prospectively randomized clinical trials have established the efficacy of strontium-89 as a first-line therapy and as an adjuvant to external-beam radiotherapy. Particularly advantageous is its usefulness in situations in which external-beam radiotherapy options have been exhausted and normal tissue tolerance has been reached. Newer radiopharmaceuticals are still under investigation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
459984
Rhenium, foil, thickness 1.0 mm, 99.98% trace metals basis
Re
267317
Rhenium, foil, thickness 0.25 mm, 99.98% trace metals basis
Re
204188
Rhenium, powder, 99.995% trace metals basis
Re
267279
Rhenium, powder, −100 mesh, ≥99.9% trace metals basis
Re
GF57863931
Rhenium, wire reel, 0.1m, diameter 0.25mm, annealed, 99.97%
Re
GF60560513
Rhenium, wire reel, 0.1m, diameter 0.178mm, annealed, 99.97%
Re
GF90374441
Rhenium, wire reel, 0.1m, diameter 0.3mm, annealed, 99.97%
Re
GF77806367
Rhenium, wire reel, 0.1m, diameter 0.5mm, annealed, 99.97%
Re
GF63991929
Rhenium, wire reel, 0.1m, diameter 1.0mm, annealed, 99.97%
Re
GF14519156
Rhenium, wire reel, 0.2m, diameter 0.076mm, annealed, 99.97%
Re
GF59830139
Rhenium, wire reel, 0.2m, diameter 0.10mm, annealed, 99.97%
Re
GF96591875
Rhenium, wire reel, 0.2m, diameter 0.125mm, annealed, 99.97%
Re
GF02183217
Rhenium, wire reel, 0.2m, diameter 0.15mm, annealed, 99.97%
Re
GF70064658
Rhenium, wire reel, 0.2m, diameter 0.178mm, annealed, 99.97%
Re
GF32991486
Rhenium, wire reel, 0.2m, diameter 0.25mm, annealed, 99.97%
Re
GF48358435
Rhenium, wire reel, 0.2m, diameter 0.2mm, annealed, 99.97%
Re
GF96805884
Rhenium, wire reel, 0.2m, diameter 0.3mm, annealed, 99.97%
Re
GF25796010
Rhenium, wire reel, 0.2m, diameter 0.5mm, annealed, 99.97%
Re
GF98990050
Rhenium, wire reel, 0.2m, diameter 1.0mm, annealed, 99.97%
Re
GF14894163
Rhenium, wire reel, 0.5m, diameter 0.076mm, annealed, 99.97%
Re
GF78766745
Rhenium, wire reel, 0.5m, diameter 0.10mm, annealed, 99.97%
Re
GF73984797
Rhenium, wire reel, 0.5m, diameter 0.125mm, annealed, 99.97%
Re
GF72901324
Rhenium, wire reel, 0.5m, diameter 0.15mm, annealed, 99.97%
Re
GF58081945
Rhenium, wire reel, 0.5m, diameter 0.178mm, annealed, 99.97%
Re
GF23936104
Rhenium, wire reel, 0.5m, diameter 0.25mm, annealed, 99.97%
Re
GF44804325
Rhenium, wire reel, 0.5m, diameter 0.2mm, annealed, 99.97%
Re
GF00735353
Rhenium, wire reel, 0.5m, diameter 0.3mm, annealed, 99.97%
Re
GF24502115
Rhenium, wire reel, 0.5m, diameter 0.5mm, annealed, 99.97%
Re
GF61770225
Rhenium, wire reel, 1m, diameter 0.076mm, annealed, 99.97%
Re
GF45842733
Rhenium, wire reel, 1m, diameter 0.10mm, annealed, 99.97%
Re