EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Alteration in the tranquilizing potency of chlorpromazine in rats exposed chronically to the insecticide, endosulfan.
PMID 7833600
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G1128 Tragacanth