EMAIL THIS PAGE TO A FRIEND
Genomics
The retinal outer segment membrane protein-1 gene (Rom1) maps to the proximal end of mouse chromosome 19.
PMID 7835908
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV44260
Anti-ROM1 antibody produced in rabbit, affinity isolated antibody