EMAIL THIS PAGE TO A FRIEND
BioTechniques
Automated procedures for the quantitation of protein.
PMID 7840981
Abstract
An entirely automated 96-well microplate-based system to perform procedures for the measurement of protein is described. This single instrument system utilizes a series of computer-controlled mechanical subsystems to move the plate, control incubations and dispense samples or reagents in order to perform the assay. This system allows the investigator to reproducibly perform these protein assays on large numbers of biological samples with minimal effort.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
15270
Biuret, 98%
C2H5N3O2