EMAIL THIS PAGE TO A FRIEND
Yan ke xue bao = Eye science
Delayed hyphema after posterior chamber intraocular lens implantation.
PMID 7843383
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5877
Adrenochrome semicarbazone
C10H12N4O3