EMAIL THIS PAGE TO A FRIEND
Deutsche medizinische Wochenschrift (1946)
[Monoethylglycinexylidide (MEGX)-test. A test for the assessment of prognosis before and after liver transplantation].
PMID 7851289
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0087
MEGX, ≥95% (HPLC)
C12H18N2O