EMAIL THIS PAGE TO A FRIEND
FEBS letters
Quinacrine mustard and lipophilic cations inhibitory to both vacuolar H(+)-ATPase and F0F1-ATP synthase.
PMID 7851533
Abstract
Various lipophilic cations, such as quinacrine mustard and dequalinium, which are known to inhibit mitochondrial F1-ATPase, strongly inhibited vacuolar H(+)-ATPase purified from bovine adrenal chromaffin granules. Quinacrine mustard bound irreversibly to vacuolar H(+)-ATPase subunit A, and the 115 kDa accessory polypeptide and dithiothreitol had no effect. The binding was competitively inhibited by chlorpromazine and quinacrine, and these compounds specifically reduced the amount of labeling of subunit A. Quinacrine mustard also prevented the binding of [alpha-32P]ATP to subunit A but had no effect on the binding of [3H]N-ethylmaleimide to either subunit A or the 115 kDa accessory polypeptide. These results suggest that the binding site of quinacrine mustard in subunit A is not related to the N-ethylmaleimide-binding site(s), which is important for activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Q2876
Quinacrine mustard dihydrochloride, ≥85% (HPLC)
C23H28Cl3N3O · 2HCl