EMAIL THIS PAGE TO A FRIEND
Annual review of biochemistry
Concepts and perspectives in enzyme stereochemistry.
PMID 786152
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
43381
(S)-Sodium 2,3-dihydroxyisovalerate hydrate, ≥95.0% (CE)
C5H10O4