EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Biomedical applications
Simultaneous measurement of nicotinic acid and its major metabolite, nicotinuric acid in urine using high-performance liquid chromatography: application of solid-liquid extraction.
PMID 7866544
Abstract
The concentrations of nicotinic acid (NiAc) and nicotinuric acid (NiUAc), a major metabolite of NiAc, were simultaneously determined in urine using solid-phase extraction (cation-exchange extraction) and reversed-phase high-performance liquid chromatography with ultraviolet detection. The intra- and inter-day precision studies showed good reproducibilities: the coefficients of variations were less than 8.1% for NiAc and 8.8% for NiUAc. The calibration curves were linear (r2 > 0.9934) in the concentration range 10-1000 micrograms/ml. The removal of endogenous interferences in urine by solid-phase extraction presented here is superior to the pretreatment protocols reported previously by other workers. The method was used in a preliminary pharmacokinetic study in rats after intravenous administration of NiAc (5 and 15 mg/kg).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N4751
Nicotinuric acid
C8H8N2O3