EMAIL THIS PAGE TO A FRIEND
Archives of biochemistry and biophysics
The inactivation of bifunctional peptidylglycine alpha-amidating enzyme by benzylhydrazine: evidence that the two enzyme-bound copper atoms are nonequivalent.
PMID 7872809
Abstract
Peptidylglycine alpha-amidating enzyme catalyzes the two-step conversion of C-terminal glycine-extended peptides to C-terminal alpha-amidated peptides and glyoxylate in a reaction that requires O2, ascorbate and 2 mol of copper per mole of enzyme [Kulathila et al. (1994) Arch. Biochem. Biophys. 311, 191-195]. Peptides with a C-terminal alpha-hydroxyglycine residue are intermediates in the amidation reaction. Benzylhydrazine inactivates the enzymatic conversion of dansyl-Tyr-Val-Gly to dansyl-Tyr-Val-NH2 in a time- and concentration-dependent manner. In contrast, the enzymatic conversion of dansyl-Tyr-Val-alpha-hydroxyglycine to dansyl-Tyr-Val-NH2 is unaffected by benzylhydrazine. The plot of 1/(inactivation rate) vs 1/[benzylhydrazine] is parabolic, indicating that the inactivation results from the interaction of 2 mol of benzylhydrazine per mole of enzyme. EPR spectra obtained from benzylhydrazine inactivation reactions carried out in the presence of a radical trap, alpha-(4-pyridyl-1-oxide)-N-tert-butylnitrone, show the formation of a carbon-centered benzyl radical. The benzyl radical most likely results from redox chemistry between benzylhydrazine and the enzyme-bound Cu(II) ions because EPR studies show that enzyme-bound Cu(II) is reduced to Cu(I) in the presence of benzylhydrazine. The kinetic constants for benzylhydrazine as a reductant in the amidation reaction were determined at benzylhydrazine concentrations too low to cause significant enzyme inactivation. Mimosine exhibits mixed inhibition vs benzylhydrazine; however, previous results have shown that benzylhydrazine is competitive vs ascorbate [Miller et al. (1992) Arch. Biochem. Biophys. 298, 380-388]. This change in kinetic mechanism coupled with the nonlinear inactivation kinetics have lead to a proposal that the two enzyme-bound Cu(II) atoms are nonequivalent with respect to their reduction by benzylhydrazine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B22852
Benzylhydrazine dihydrochloride, 97%
C7H10N2 · 2HCl