EMAIL THIS PAGE TO A FRIEND
Journal of neurology, neurosurgery, and psychiatry
Pholedrine: a substitute for hydroxyamphetamine as a diagnostic eyedrop test in Horner's syndrome.
PMID 7876854
Abstract
Mydriatic responses to eyedrops containing the indirect acting sympathomimetic amines tyramine, hydroxyamphetamine, and pholedrine have been compared in 10 healthy subjects. Pholedrine, the n-methyl derivative of hydroxyamphetamine, at a concentration of 1% had effects similar to those produced by 0.5% hydroxyamphetamine itself. Pretreatment with topical guanethidine attenuated its responses and in 13 patients with unilateral Horner's syndrome it distinguished clearly those five patients who had preganglionic from the eight with postganglionic lesions. It is concluded that 1% pholedrine may be substituted for 0.5% hydroxyamphetamine, which is no longer available, as a diagnostic agent for use in Horner's syndrome.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H9645
p-Hydroxymethamphetamine, analytical standard, for drug analysis
C10H15NO