EMAIL THIS PAGE TO A FRIEND
World journal of urology
Biometric assessment of prostate cancer's metastatic potential.
PMID 7881466
Abstract
Currently, no protocol exists that can assess the metastatic potential of prostate adenocarcinoma. The reason for this is partly due to the lack of information on cellular changes that result in a tumor cell's becoming metastatic. In this investigation, attempts were made to devise a method that correlated with the metastatic potential of AT-1, Mat-Lu, and Mat-LyLu cell lines of the Dunning R-3327 rat prostatic adenocarcinoma system. To accomplish this, we applied BioQuant biometric parameters, i.e., area, shape factor, and cell motility. AT-1 had a lower shape factor and a greater area as compared with the more highly metastatic Mat-Lu subline. No significant difference in area or shape factor was detected between the AT-1 cell line and the highly metastatic Mat-LyLu line. However, the lowly metastatic AT-1 line had less motility as compared with the Mat-Lu and Mat-LyLu lines. This study revealed that metastatic potential could be partially predicted via area and shape factor and accurately predicted via cell motility.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
AT-1, 94101449
details
MAT-LyLu, 94101454