EMAIL THIS PAGE TO A FRIEND
Yao xue xue bao = Acta pharmaceutica Sinica
[Antagonism of daurisoline on arrhythmogenic delayed afterdepolarization].
PMID 7900534
Abstract
Standard microelectrode technique was used to study the effect of daurisoline (DS) on delayed afterdepolarization (DAD) and triggered activity (TA) in guinea pig ventricular trabeculae. DS (50 mumol.L-1) abolished TA induced by ouabain or caffeine, inhibited isoprenaline-induced TA, decreased incidence of TA induced by K(+)-free and high-Ca2+ medium or phenylephrine from 100.75% to 9.1% and 22.2% respectively, and significantly suppressed DAD caused by the above induces. It is concluded that DS exert antagonistic action to DAD and TA in guinea pig trabeculae.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0597 Daurisoline, ≥98% (HPLC)
C37H42N2O6