EMAIL THIS PAGE TO A FRIEND
Neuropharmacology
(2S,3S,4R)-2-(carboxycyclopropyl)glycine, a potent and competitive inhibitor of both glial and neuronal uptake of glutamate.
PMID 7901789
Abstract
The effects of several diastereoisomers of L-2-(carboxycyclopropyl)glycine (CCG) on L-glutamate uptake were compared among three different preparations, glial plasmalemmal vesicles (GPV), synaptosomes and cultured astrocytes from rat hippocampus. The (2S,3S,4R)-isomer (L-CCG-III) inhibited a Na(+)-dependent high-affinity L-glutamate uptake in GPV and synaptosomes in a dose dependent manner at a micromolar range. The potency was quite similar to that of L-threo-beta-hydroxyaspartate in both subcellular fractions and much higher than L-aspartate-beta-hydroxamate, which were known as potent inhibitors of glutamate uptake. The (2S,3R,4S)-isomer (L-CCG-IV) also inhibited the glutamate uptake in GPV and synaptosomes, but it was about 100 times less active than L-CCG-III. The (2S,3S,4S)- and (2S,3R,4R)-isomers (L-CCG-I and L-CCG-II, respectively) hardly showed any inhibitory action on the glutamate uptake. Dixon plot analysis of the initial uptake rate revealed that the inhibition was in a competitive manner and the value of the inhibition constant (Ki) was about 1 microM in both GPV and synaptosomes. L-CCG-III effectively inhibited the glutamate uptake by cultured hippocampal astrocytes as well. These results suggested that L-CCG-III inhibited the glutamate uptake in both neurones and glial cells of the mammalian central nervous system in a similar manner.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A6508
L-Aspartic acid β-hydroxamate, serine racemase inhibitor
C4H8N2O4