EMAIL THIS PAGE TO A FRIEND
Carcinogenesis
Enhancement of altered hepatic foci in rat liver and inhibition of intercellular communication in vitro by the pyrethroid insecticides fenvalerate, flucythrinate and cypermethrin.
PMID 7903603
Abstract
Male Sprague-Dawley rats dosed with N-nitrosodiethylamine (NDEA) 24 h after two-thirds partial hepatectomy were treated with the pyrethroid insecticides fenvalerate, flucythrinate or cypermethrin in the diet for 20 weeks. Altered hepatic foci were analyzed by quantitative stereology from paraffin-embedded sections stained for gamma-glutamyltranspeptidase (GGT) or glutathione S-transferase P (GST-P). The present results demonstrate that the pyrethroids tested all enhance the development of NDEA-initiated, GGT-positive foci in rat liver at non-hepatotoxic doses. On the contrary, the volume fractions of GST-P-positive foci were not elevated as compared to the control group. The three pyrethroids tested all inhibited the transfer of Lucifer Yellow CH between WB-F344 rat liver epithelial cells in culture, supporting the increase of GGT-positive foci and suggesting that these substances can act as tumour promoters. The discrepancy between the results from analyses using GGT or GST-P as markers emphasizes the importance of understanding the mechanism underlying the expression of different markers for preneoplastic lesions and the importance of such effects in tumour promotion.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33496
Flucythrinate, PESTANAL®, analytical standard
C26H23F2NO4