EMAIL THIS PAGE TO A FRIEND
Journal of the National Cancer Institute
A 190-kilodalton protein overexpressed in non-P-glycoprotein-containing multidrug-resistant cells and its relationship to the MRP gene.
PMID 7903701
Abstract
A 190k (190-kilodalton) membrane protein has been identified in several multidrug-resistant (MDR) cell lines that show decreased drug accumulation without expression of P-glycoprotein. It is not clear whether this 190k protein is involved directly in drug efflux. Recently, a gene for a putative transporter protein, MRP (multidrug resistance-associated protein) has been sequenced and localized to chromosome 16. The protein encoded by this gene contains a 7-amino-acid sequence present in the synthetic peptide used to generate the antiserum recognizing the 190k protein. The study was undertaken to clarify the relationship of the 190k protein to MRP gene expression in non-P-glycoprotein-containing MDR cells of the large-cell and adenocarcinoma lung cancer lines, COR-L23 and MOR. Expression of the 190k protein was determined by Western blot analysis and that of the MRP gene by polymerase chain reaction amplification of complementary DNA reverse transcribed from RNA. Abnormalities of chromosome 16 were investigated in chromosome spreads by fluorescence in situ hybridization. The amount of detectable 190k protein is closely associated with degree of drug resistance. Cell lines surviving in higher drug concentrations have greater amounts of protein, and revertant lines grown without drug for up to 28 weeks show reduced expression of the protein together with enhanced drug sensitivity. The 190k protein appears to be one of the major proteins differentially expressed in membranes of drug-resistant cells. The amount of MRP messenger RNA correlates closely with that of the 190k protein. The MDR cells contain amplified chromosome 16 material with many double minutes in the large-cell lung tumor lines and an enlarged chromosome 16 in the adenocarcinoma lines. The 190k protein detected immunologically is likely to be the protein, encoded by the MRP gene, which becomes overexpressed in these cells as a consequence of chromosomal amplification and fragmentation. Though associated with drug resistance, enhanced drug efflux, and decreased drug accumulation in cell lines, the role of this protein in clinical resistance has yet to be determined.