EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical sciences
Metabolites of ebrotidine, a new H2-receptor antagonist, in human urine.
PMID 7909553
Abstract
Ebrotidine is a new H2-receptor antagonist which exhibits a remarkable ability for gastric mucosal protection. A preliminary metabolic pathway for this compound was proposed and the hypothetic metabolites were synthesized. The presence of ebrotidine and its metabolites ebrotidine S-oxide and 4-bromobenzenesulfonamide in human urine has been confirmed by HPLC separation and spectroscopic characterization of the collected fractions by FT-IR and 1H NMR. Ebrotidine S,S-dioxide has been identified by HPLC using diode-array detection.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
16374
4-Bromobenzenesulfonamide, ≥99.0% (T)
C6H6BrNO2S