EMAIL THIS PAGE TO A FRIEND
The Journal of pharmacy and pharmacology
A stable and highly potent hexamethonium derivative which modulates muscarinic receptors allosterically in guinea-pig hearts.
PMID 7912726
Abstract
W84 (N,N,N',N'-tetramethyl-N,N'-bis-(3-phthalimidopropyl)-N,N'-hexane- 1,6- diyl-bis-ammonium dibromide) is a potent stabilizer of antagonist binding to muscarinic receptors; however, W84 hydrolyses in aqueous buffered medium. The synthesis of the stable derivative CHIN3/6 is presented containing a 2-phenyl-quinazolinone instead of the labile phthalimide substituent. This compound retarded [3H]N-methylscopolamine-dissociation in guinea-pig cardiac membranes with slightly higher potency than W84, the EC50 values amounting to 7.5 x 10(-7) and 13 x 10(-7) M, respectively. Molecular modelling revealed differences in the electron density of the substituents and in their molecular shape. It is suggested that the derivatives use partially different sites of attachment when occupying the allosteric binding site of the receptor protein.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W0641
W-84 dibromide, ≥98% (HPLC), solid
C32H44Br2N4O4