EMAIL THIS PAGE TO A FRIEND
Clinical science (London, England : 1979)
Renal effects of local infusion of adenosine in man.
PMID 7924159
Abstract
1. The effects of local intra-arterial infusion of adenosine on renal blood flow, glomerular filtration and renin release in eight healthy awake subjects have been examined. 2. Renal blood flow and glomerular filtration rate were measured as the clearances of p-aminohippurate and inulin, respectively. After basal samplings, adenosine was infused intra-arterially at successive rates of 2 and 10 micrograms min-1 kg-1 for 40 min at each rate. 3. Apart from a small increase in heart rate (65 +/- 4 to 71 +/- 4 beats/min), there were no signs of sympathetic activation (unchanged blood pressure and catecholamine levels) during the infusion. Clearance of p-aminohippurate tended to increase, but not significantly, during adenosine infusion (518 +/- 48 ml/min basal, 563 +/- 52 ml/min during the highest dose of adenosine). The arterial plasma concentration of p-aminohippurate decreased by 9 +/- 3% (P < 0.05), consistent with a small increase in renal blood flow in the infused kidney. Inulin clearance was reduced from 115 +/- 3 to 97 +/- 2 ml/min (P < 0.001). The extraction of inulin, reflecting the filtration fraction, was 18% in both kidneys in the basal state. During infusion of adenosine the extraction in the infused kidney decreased to 12 +/- 3% (P < 0.01 compared with the control kidney, 23 +/- 3%). 4. The total excretion of Na+ was unchanged, but there was a minor decrease in K+ clearance. Thus, the K+/Na+ excretion ratio decreased from a basal value of 13 +/- 2 to 10 +/- 2 (P < 0.01) at the highest dose of adenosine.(ABSTRACT TRUNCATED AT 250 WORDS)
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1422
p-Aminohippuric acid, ≥99%
C9H10N2O3
08090
p-Aminohippuric acid, ≥98.0% (T)
C9H10N2O3