EMAIL THIS PAGE TO A FRIEND
The Journal of antibiotics
Novel carbon-carbon bond cleavage reaction of penem antibiotic by class C beta-lactamases.
PMID 7928684
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F8182
Faropenem sodium hydrate, ≥98% (HPLC)
C12H14NNaO5S · xH2O