EMAIL THIS PAGE TO A FRIEND
Journal of neural transmission. Supplementum
Amphetamine-metabolites of deprenyl involved in protection against neurotoxicity induced by MPTP and 2'-methyl-MPTP.
PMID 7931228
Abstract
The ability of 1-deprenyl to protect against the parkinsonian effects of 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP) has been attributed to the inhibition of conversion of MPTP to MPP+ (1-methyl-4-phenylpyridinium) catalyzed by MAO-B. We report here that deprenyl-treatment in mice has an additional neuroprotective element associated with the rapid metabolization of 1-deprenyl to 1-methamphetamine and 1-amphetamine. 1-Methamphetamine and 1-amphetamine inhibit MPP(+)-uptake into striatal synaptosomes prepared from rats. Post-treatment by 1-deprenyl, 1-methamphetamine, 1-amphetamine (at times when MPTP is no longer present in the striatum of mice) protects against neurotoxicity in C57BL mice by blocking the uptake of MPP+ into dopaminergic neurons, and even against the neurotoxicity induced by 2'CH3-MPTP, which is partly bioactivated by MAO-A. These findings may have clinical implications since deprenyl has recently been found to delay the progression of Parkinson's disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M103
1-Methyl-4-(2′-methylphenyl)-1,2,3,6-tetrahydropyridine hydrochloride, solid
C13H17N · HCl