EMAIL THIS PAGE TO A FRIEND
JAMA
Examining product risk in context: the case of zomepirac.
PMID 7933364
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Z2625
Zomepirac sodium salt
C15H13ClNNaO3