EMAIL THIS PAGE TO A FRIEND
Anti-cancer drugs
Antimetastatic effects of synthetic peptides containing the core sequence of the type III connecting segment domain (IIICS) of fibronectin.
PMID 7949246
Abstract
The antimetastatic activities of synthetic peptides corresponding to fragments of the adhesion-related molecules, such as fibronectin and laminin, were examined. We prepared three peptides derived from the type III connecting segment domain (IIICS) of fibronectin: Glu-Ile-Leu-Asp-Val (EILDV), Glu-Ile-Leu-Asp-Val-Pro-Ser-Thr (EILDVPST), Arg-Glu-Asp-Val (REDV), and a laminin-related peptide, Tyr-Ile-Gly-Ser-Arg (YIGSR). Each peptide inhibited the experimental tumor metastasis of B16-BL6 melanoma, while EILDV had the strongest effect. The peptides conjugated with poly(ethylene glycol) (PEG) were more effective than the unmodified peptides in molar ratio terms. A mixture composed of PEG hybrids with EILDV, REDV and YIGSR significantly inhibited tumor metastasis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F6398
Fibronectin Type III Connecting Segment Fragment 90-109, ≥97% (HPLC)
C107H158N28O34