EMAIL THIS PAGE TO A FRIEND
Receptor
Evidence for chemical differentiation of delta opioid receptor subtypes by the sulfhydryl reagent N-ethylmaleimide.
PMID 7950982
Abstract
In this study, the delta receptor-selective nonequilibrium affinity ligands, 5'-NTII and DALCE, and the nonspecific sulfhydryl reagent NEM were evaluated over a range of concentrations and treatment conditions for their ability to selectively alter the binding properties of delta 1- or delta 2-preferring opioid radioligands in brain homogenate. Treatment of tissue preparations with DALCE (0-10,000 nM) or NTII (0-10,000 nM) resulted in an equivalent concentration-dependent loss of binding capacity for the delta 1 agonist 3H-DPDPE and the mu/delta 2 agonist 3H-DSLET. In contrast, treatment of tissue with NEM (0-8000 microM) resulted in greater loss of 3H-DPDPE binding. Scatchard analysis of the binding of 3H-DPDPE, 3H-DSLET, and 3H-NTI in 3 mM NEM-treated rat brain P2 preparation revealed an equivalent decrease in affinity for the agonist ligands, but a significantly greater decrease in Bmax for 3H-DPDPE compared with control tissue values. Comparison of the K(i) values for a series of delta-selective compounds against 3H-DSLET binding in control vs 3 mM NEM treated P2 fraction showed differential effects of NEM on affinity within the series that were consistent with a selective depletion of delta 1 sites. Overall, these results indicate that NEM treatment selectively reduced delta 1 receptor binding, resulting in a preparation that is enriched in delta 2 sites.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N157
Naltrindole isothiocyanate hydrochloride, solid
C27H25N3O3S · HCl