EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography. B, Biomedical Applications
Assay for maprotiline in human serum with improved sensitivity and selectivity.
PMID 7952036
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
13-111
MBP, biotin conjugate