EMAIL THIS PAGE TO A FRIEND
Journal of virology
Varicella-zoster virus (VZV) open reading frame 10 protein, the homolog of the essential herpes simplex virus protein VP16, is dispensable for VZV replication in vitro.
PMID 7966575