EMAIL THIS PAGE TO A FRIEND
Sudebno-meditsinskaia ekspertiza
[The characteristics of extracting 2-acetylamino-5-nitrothiazole and 5-nitro-8-hydroxyquinoline from aqueous solutions].
PMID 7974608
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
140325
8-Hydroxy-5-nitroquinoline, 96%
C9H6N2O3