EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Measurement of excitatory sulfur amino acids, cysteine sulfinic acid, cysteic acid, homocysteine sulfinic acid, and homocysteic acid in serum by stable isotope dilution gas chromatography-mass spectrometry and selected ion monitoring.
PMID 7978266
Abstract
Oxidized sulfur-containing amino acids are recognized as agonists of excitatory amino acid receptors in the mammalian nervous system. Homologues of glutamic acid (homocysteine sulfinic acid and homocysteic acid) and aspartic acid (cysteine sulfinic acid and cysteic acid) have been shown to be agonistic to N-methyl-D-aspartate receptors in animal brain and have been demonstrated in brain tissue. Considerable evidence exists for the role of homocysteic acid and cysteine sulfinic acid as endogenous ligands for excitatory amino acid receptors. We report, for the first time, the quantitation of these compounds in normal human serum, by a newly developed gas chromatography-mass spectrometry method that employs stable isotope-dilution selected ion monitoring using internal standards prepared in our laboratory. We also report new methods of synthesis of stable isotope-labeled internal standards used in measuring cysteine sulfinic acid, cysteic acid, homocysteine sulfinic acid, and homocysteic acid.