EMAIL THIS PAGE TO A FRIEND
Comparative biochemistry and physiology. Pharmacology, toxicology and endocrinology
Insecticidal and cytotoxic effects of natural and hemisynthetic destruxins.
PMID 7981981
Abstract
The insecticidal and cytotoxic effects of 13 natural and hemisynthetic destruxins have been studied. DE shows insecticidal effects similar to those of DA, while DE and DA are more active than all the other natural compounds and analogues tested. Brominated destruxin is a relatively active analogue displaying particular modalities of cytotoxic effects which reflect a certain originality of its mode of action. The linear molecule resulting from the opening of the DA cycle is not toxic. The most hydrophilic destruxins showing e.g. charged radicals (COO-) appear the least toxic probably because they do not penetrate easily the cellular membranes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D4921
Destruxin A from Metarrhizium anisopliae, ≥98% (HPLC)
C29H47N5O7