EMAIL THIS PAGE TO A FRIEND
Microvascular research
In situ study of the membrane potential in microvascular endothelial cells using a fluorescent probe.
PMID 7990720
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
44977
DISBAC2(3), Voltage Sensitive Probe, ≥98% (HPLC)
C19H24N4O4S2