EMAIL THIS PAGE TO A FRIEND
Trends in pharmacological sciences
Novel receptor antagonists welcome a new era in endothelin biology.
PMID 7992380
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B150
BQ-123, ≥99%, sodium salt, lyophilized powder
C31H41N6O7 Na