EMAIL THIS PAGE TO A FRIEND
Bioorganicheskaia khimiia
[Synthesis and biological activity of new analogs of beta-casomorphine-5].
PMID 7993378
Abstract
Four new analogues of beta-cazomorphine-5 modified at the C-end with ethylenediamine- and glycine-containing derivatives were synthesized by the standard method of peptide chemistry (mixed anhydrides, carbodiimide, activated esters): H-Tyr-Pro-Phe-Pro-Gly-EtDA-Gly-II (I) H-Tyr-Pro-Phe-Pro-Gly-EtDA-Gly-Gly-II (II) H-Tyr-Pro-Phe-Pro-Gly-EtDA-Gly-Gly-CO-CH3 (III) H-Tyr-Pro-Phe-Pro-Gly-EtDA-Gly-Gly-CO-CH2-CH2-COOH (IV) The level of affinity and the degree of selectivity of the peptides towards the mu- and delta-opioid receptors of the rat brain lyophilized membranes were studied by the radioreceptor method. All the new peptides displayed analgetic activity, largely depending upon their structure.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C5147
β-Casomorphin Fragment 1-5 hydrochloride, ≥97% (HPLC)
C30H37N5O7