EMAIL THIS PAGE TO A FRIEND
Rapid communications in mass spectrometry : RCM
Charge-remote fragmentation of peptides derivatized with 4-aminonaphthalenesulphonic acid.
PMID 8000077
Abstract
A series of small peptides has been studied by negative-ion fast-atom bombardment mass spectrometry with collision-induced dissociation. It has been found that by derivatizing peptides with 4-aminonaphthalenesulphonic acid in a peptide linkage at the C-terminus, negative-ion formation can be enhanced and fragmentation in collision-induced dissociation reactions controlled. The peptide-naphthalenesulphonates show charge-remote fragmentations and the resultant spectra give sequence information.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
250619
4-Amino-1-naphthalenesulfonic acid, 97%
C10H9NO3S