EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Survey of pesticide use by Ontario greenhouse chrysanthemum producers.
PMID 8000174
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45443
Dienochlor, PESTANAL®, analytical standard
C10Cl10
N12741 Oxydemeton-methyl, analytical standard
C6H15O4PS2