EMAIL THIS PAGE TO A FRIEND
Acta crystallographica. Section C, Crystal structure communications
Non-steroidal anti-inflammatory drugs. III. Structure of indoprofen: 2-[4-(1-oxo-2-isoindolinyl)phenyl]propionic acid.
PMID 8011271
Abstract
Indoprofen, 4-(1,3-dihydro-1-oxo-2H-isoindol-2-yl)-alpha-methylbenzeneaceti c acid, C17H15NO3, a non-steroidal anti-inflammatory agent, falls into the class of phenylpropionic acids that includes ibuprofen, ketoprofen and flubirprofen. It adopts a planar conformation apart from the carboxylic acid group. There is a small twist of 8.8 (2) degrees between the mean planes of the isoindolinyl and phenyl rings. The rotation of the carboxyl group out of the plane of the adjacent phenyl ring seems to be connected with anti-inflammatory activity and is similar to that found in other 2-phenylpropionic acids. A pair of intermolecular hydrogen bonds connects two carboxylic acid groups across a centre of symmetry with an O...O distance of 2.61 (1) A.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I3132
Indoprofen, analytical standard
C17H15NO3