EMAIL THIS PAGE TO A FRIEND
Journal of AOAC International
Rapid abrasion test to indicate lead on the surface of ceramicware.
PMID 8012226
Abstract
A rapid abrasion test (RAT) was developed for screening ceramicware for Pb. A Pb-sensitive chromogen (rhodizonic acid) reacts to form a persistent colored complex, indicating the presence of Pb. RAT takes 2-5 min to complete; a positive test is easily discernible. RAT, which provides only qualitative information, is useful for screening large numbers of ceramicware items for the presence of Pb at > or = 0.05 microgram/mL.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
392340
Potassium rhodizonate, technical grade, 85%
C6K2O6